Learn More
Texas Medical Industries SAF-EVAC FOR ISTAT 100/PK
SDP
Catalog No. NC0450377
Encompass
Saf-Evac For Use With Your I-Stat Device. Economical Tool That Limits Technologists Exposure To Etiological Agents In Blood Samples. Saf-Evac Prevents The Aerosolization (Splashing) Of Blood Normally Associated With Uncapping Of Evacuated Sampling Tubes. The Device Allows Laboratory Personnel To Dispense Whole Anti Coagulated Blood From The Tube One Drop At A Time. The Blood Smear May Now Be Prepared Without Removal Of The Stopper.
Catalog No. NC0450377 Supplier Texas Medical Industries Supplier No. 5002
Saf-Evac For Use With Your I-Stat Device. Economical Tool That Limits Technologists Exposure To Etiological Agents In Blood Samples. Saf-Evac Prevents The Aerosolization (Splashing) Of Blood Normally Associated With Uncapping Of Evacuated Sampling Tubes. The Device Allows Laboratory Personnel To Dispense Whole Anti Coagulated Blood From The Tube One Drop At A Time. The Blood Smear May Now Be Prepared Without Removal Of The Stopper.