Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Safari Family Reunion with SYBR™ Safe
SDPSCI_ED

Catalog No. 01672277 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
01-672-277 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-277 Supplier EDVOTEK™ Supplier No. 920S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this Safari Family Reunion with SYBR™ Safe lab, students will perform electrophoresis on the DNA samples of two lions in order to return them to wildlife sanctuaries close to their ancestral home.

  • Includes SYBR™ Safe DNA Stain, NOT FlashBlue™ DNA Stain
  • Learn how DNA is used to discover an individual's ancestry
  • Perform DNA electrophoresis and RFLP analysis
  • Analyze phylogenetic tree and haplotype maps
  • Explore how conservation biologists use genetic data
  • Group Size: For 8 Gels
  • Time Required: Complete in 45 minutes
  • Kit Includes: Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), SYBR™ Safe DNA Stain, Practice Gel Loading Solution.
  • Storage: Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.

Specifications

Grade 8 to Undergraduate
Class Size 8 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.