Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Safari Reunion
SDPSCI_ED

Catalog No. S52003 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S52003 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S52003 Supplier Edvotek™ Supplier No. 920
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this lab, students will perform electrophoresis on the DNA samples of two lions in order to return them to wildlife sanctuaries close to their ancestral home.

  • Learn how DNA is used to discover an individual's ancestry
  • Perform DNA electrophoresis and RFLP analysis
  • Analyze phylogenetic tree and haplotype maps
  • Explore how conservation biologists use genetic data
  • Designed for 8 gels if stained with FlashBlue™ (included) or 16 gels if stained with SYBR™ Safe (not included)
  • Complete gel electrophoresis in 45 minutes and analysis in 20-30 minutes
  • Kit includes Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), Practice Gel Loading Solution, FlashBlue™ DNA Stain
  • Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended

Specifications

Grade 8 to Undergraduate
Age 14 to 20
For Use With (Application) Perform DNA electrophoresis and RFLP analysis
Includes Instructions, Ready-to-Load QuickStripTM DNA Samples, UltraSpec-AgaroseTM, Electrophoresis Buffer (50 X), Practice Gel Loading Solution, FlashBlueTM DNA Stain
Requires DNA Electrophoresis, Micropipettes: 5-50 μL (Optional), White Light Box, & Microwave or Hot Plate.
Content And Storage Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.
Class Size 8 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.