Learn More
Enterprise Technology Solutions Safco Commercial Shelving Kit, 5-Shelf, 36"x18"x75", D.Gray
SDP
Catalog No. 5000791147
Encompass_Preferred
Commercial steel shelving features a box beam design and double-sided compression clips to support heavy loads. Evenly loaded corner brackets help prevent rocking and greatly improve stability and rigidity. Hat channel underneath each shelf increases overall strength by helping prevent bowing and deflection from overloading. Powder-coat finish offers durability. Shelf height adjusts in 1" increments. 24-gauge steel shelf holds up to 700 lb. evenly distributed.
Catalog No. 50-007-91147 Supplier Enterprise Technology Solutions Supplier No. SAF6266
Commercial steel shelving features a box beam design and double-sided compression clips to support heavy loads. Evenly loaded corner brackets help prevent rocking and greatly improve stability and rigidity. Hat channel underneath each shelf increases overall strength by helping prevent bowing and deflection from overloading. Powder-coat finish offers durability. Shelf height adjusts in 1" increments. 24-gauge steel shelf holds up to 700 lb. evenly distributed.