Learn More
Enterprise Technology Solutions Safco Starter Shelving Unit,4 Shelves/Posts,800 lb,48"x24"x72",BK
SDP
Catalog No. 5000791415
Encompass_Preferred
Industrial wire shelving offers a dust-free wire design and adjustable leveling feet with plastic caps to protect floors. Flexible design allows 1" shelf adjustments and modular expansion. Each shelf holds 800 lb. evenly distributed. Wire shelving is made from strong, welded steel wire construction (10-gauge shelf surface, 3-gauge frame, 16-gauge posts, 6-gauge wave pattern) with a durable powder-coat finish. Open wire design permits air circulation and prevents dust accumulation. Add-on (sold separately) and starter units can be combined to expand your shelving system. Easy collar-and-sleeve assembly requires no tools. Starter unit includes four shelves, four posts and snap-together clips. Add-on unit (sold separately) includes four shelves, two posts and snap-together clips.
Catalog No. 50-007-91415 Supplier Enterprise Technology Solutions Supplier No. SAF5294BL
Industrial wire shelving offers a dust-free wire design and adjustable leveling feet with plastic caps to protect floors. Flexible design allows 1" shelf adjustments and modular expansion. Each shelf holds 800 lb. evenly distributed. Wire shelving is made from strong, welded steel wire construction (10-gauge shelf surface, 3-gauge frame, 16-gauge posts, 6-gauge wave pattern) with a durable powder-coat finish. Open wire design permits air circulation and prevents dust accumulation. Add-on (sold separately) and starter units can be combined to expand your shelving system. Easy collar-and-sleeve assembly requires no tools. Starter unit includes four shelves, four posts and snap-together clips. Add-on unit (sold separately) includes four shelves, two posts and snap-together clips.