Learn More
Enterprise Technology Solutions Safco Waste Receptacle,Square,38 Gal, 18-1/4"x18-1/4"x31-1/2",BK
SDP
Catalog No. 5000829615
Encompass_Preferred
Indoor/outdoor waste receptacle with recessed panels features a square design and top opening. High-density polyethylene has built-in UV inhibitors to resist the harshest weather extremes. Telescoping base hides the bag. 38-gallon receptacle uses standard 32" x 44" trash bags and is easily adaptable for anchoring or weighting devices. Top opening measures 9-1/2" in diameter.
Catalog No. 50-008-29615 Supplier Enterprise Technology Solutions Supplier No. SAF9475BL
Indoor/outdoor waste receptacle with recessed panels features a square design and top opening. High-density polyethylene has built-in UV inhibitors to resist the harshest weather extremes. Telescoping base hides the bag. 38-gallon receptacle uses standard 32" x 44" trash bags and is easily adaptable for anchoring or weighting devices. Top opening measures 9-1/2" in diameter.