Learn More
Apexbio Technology LLC Safe DNA Gel Stain 100x1ml
SDP
Catalog No. 5000061916
Encompass_Preferred
Safe DNA Gel Stain is a fluorescent nucleic acid dye developed for visual detection of DNA or RNA fragments in agarose or polyacrylamide gels during electrophoresis procedures Structurally this reagent binds nucleic acids by intercalating into their double-stranded regions resulting in enhanced fluorescence upon excitation by ultraviolet (UV) or blue light transilluminators It is commonly utilized in molecular biology workflows such as agarose gel electrophoresis polyacrylamide gel electrophoresis and nucleic acid gel purification Designed as a safer alternative to ethidium bromide it reduces mutagenic risks associated with DNA staining and UV exposure thus facilitating downstream recombinant and cloning experiments Safe DNA Gel Stain is provided as a concentrated solution compatible with DNA and RNA visualization in routine laboratory gel-based analyses
Catalog No. 50-000-61916 Supplier Apexbio Technology LLC Supplier No. A8743100
Safe DNA Gel Stain is a fluorescent nucleic acid dye developed for visual detection of DNA or RNA fragments in agarose or polyacrylamide gels during electrophoresis procedures Structurally this reagent binds nucleic acids by intercalating into their double-stranded regions resulting in enhanced fluorescence upon excitation by ultraviolet (UV) or blue light transilluminators It is commonly utilized in molecular biology workflows such as agarose gel electrophoresis polyacrylamide gel electrophoresis and nucleic acid gel purification Designed as a safer alternative to ethidium bromide it reduces mutagenic risks associated with DNA staining and UV exposure thus facilitating downstream recombinant and cloning experiments Safe DNA Gel Stain is provided as a concentrated solution compatible with DNA and RNA visualization in routine laboratory gel-based analyses