Learn More
Thomas Scientific SAFE GEN3 2ML TUBES BETA 10CS
SDP
Catalog No. NC3780225
Encompass
SAFE ®GEN3, 2ml tubes, 2D bottom code, 1D & human readable side codes, pre-racked with blue caps, automation friendly rack, beta sterile, 48 tubes per rack, 10 racks per case.
Catalog No. NC3780225 Supplier Thomas Scientific Supplier No. 23A00R836
SAFE ®GEN3, 2ml tubes, 2D bottom code, 1D & human readable side codes, pre-racked with blue caps, automation friendly rack, beta sterile, 48 tubes per rack, 10 racks per case.