Learn More
Sigma Aldrich Fine Chemicals Biosciences Safe Harbor Landing Pad Ce
Catalog No. 502909061
Encompass
The STR profile of this cell line matches that of its parental cell line ATCC(R) Catalog No. TIB-152. Jurkat T lymphocytes are a human acute T cell lymphoma cell line isolated in the late 1970s from the peripheral blood of a young male patient suffering from T cell leukemia. The cells possess a pseudodiploid karyotype and have been characterized as expressing CD3 and upon stimulation interleukin-2.
Catalog No. 50-290-9061 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLL12211VL
The STR profile of this cell line matches that of its parental cell line ATCC(R) Catalog No. TIB-152. Jurkat T lymphocytes are a human acute T cell lymphoma cell line isolated in the late 1970s from the peripheral blood of a young male patient suffering from T cell leukemia. The cells possess a pseudodiploid karyotype and have been characterized as expressing CD3 and upon stimulation interleukin-2.