Learn More
Bel-Art™ Safe-Lab Thermometer Joint Adapter
Catalog No. 10029253 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Standard Taper Joint No.:
19/22
Catalog No. Standard Taper Joint No.
10-029-253 19/22
1 options
Catalog No. 10-029-253 Supplier Bel-Art™ Supplier No. F203091710
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
For tapered joints.
  • PTFE body with Viton™ O-ring for air-tight vacuum work
  • Adapter with threaded polypropylene screw cap on top
  • Inside O-ring for depth adjustment of 6 to 7mm for straight stem thermometers, glass tubing and other items
  • Rotating extracting nut removes adapter from flask
  • Not autoclavable
Specifications
Description 19/22 joint size
Includes Viton™ O-ring for air-tight vacuum work, threaded polypropylene screw cap on top, and rotating extracting nut
Material PTFE
Product Type Thermometer Joint Adapter
Standard Taper Joint No. 19/22
WARNING: Cancer - www.P65Warnings.ca.gov