Learn More
Safe Life™ N95 Respirator with Triosyn™ Antimicrobial Protection
Click to view available options
:
Universal
Description
- Designed specifically to stop airborne microorganisms including viruses
- Multi-layer system with patented Triosyn iodine-polymeric resin compound
- Captured microorganisms are rapidly oxidized and rendered harmless
- Antimicrobial protection maintains the integrity of the filtration media and prevents microbial migration and strike through over an extended period of time
- Fluid resistance protects against splashes and sprays of blood and other bodily fluids as well as wet environmental contaminants
- Activated carbon layer provides relief from organic vapors and nuisance odors
- Adjustable head straps and adjustable nose piece ensure a secure and comfortable fit
Order Info
Multiple models and sizes allow a good fit to various face types or facial features; full-surround foam gasket on models T-5000 and T-5000V facilitates snug fit on multiple face shapes. Performance is guaranteed for five years from date of manufacture, making these respirators ideal for stockpiling.
Specifications
Specifications
| Aerosol Type | N - Not Resistant to Oil |
| Certifications (NIOSH) | NIOSH 42 CFR 84, N95 |
| Design Type | Cup/Molded |
| Filter Type | N95 |
| Product Type | Healthcare Particulate Respirator and Surgical Mask |
| With Exhalation Valve | No |
| Model | T-5000 |
| FDA Cleared Surgical Mask | Yes |