Learn More
MSC Dumond Stone Cleaner, Liquid, Mild, No Scent, 1 gal
Catalog No. 502816573
Encompass
This stone cleaner is a mild, liquid formula with no scent, supplied in a 1 gal. jug. It is effective in removing various contaminants including dust, oil, grease, dirt, and ink, utilizing its water-based composition and phosphoric acid formula for cleaning efficacy.
  • Supplied in a 1 gal. jug
  • Liquid formula
  • No scent
  • Mild formula
  • Effectively removes dust, oil, grease, dirt, and ink
  • Water-based composition
  • Phosphoric acid formula
  • Manufactured in the United States
Catalog No. 50-281-6573 Supplier MSC Supplier No. 20725974
This stone cleaner is a mild, liquid formula with no scent, supplied in a 1 gal. jug. It is effective in removing various contaminants including dust, oil, grease, dirt, and ink, utilizing its water-based composition and phosphoric acid formula for cleaning efficacy.
  • Supplied in a 1 gal. jug
  • Liquid formula
  • No scent
  • Mild formula
  • Effectively removes dust, oil, grease, dirt, and ink
  • Water-based composition
  • Phosphoric acid formula
  • Manufactured in the United States