Learn More
MSC Dumond Metal Cleaner, Liquid, Phosphoric Acid, Mild, 5 gal
Catalog No. 502819088
Encompass
This liquid metal cleaner is formulated with phosphoric acid and packaged in a 5-gallon tote. It is a mild, non-scented, water-based solution specifically designed for efficient rust removal.
  • Liquid form
  • Tote container type
  • Phosphoric acid formula
  • Container size: 5.00 gal
  • Mild harshness
  • No scent
  • Removes rust
  • Water-based composition
Catalog No. 50-281-9088 Supplier MSC Supplier No. 20725750
This liquid metal cleaner is formulated with phosphoric acid and packaged in a 5-gallon tote. It is a mild, non-scented, water-based solution specifically designed for efficient rust removal.
  • Liquid form
  • Tote container type
  • Phosphoric acid formula
  • Container size: 5.00 gal
  • Mild harshness
  • No scent
  • Removes rust
  • Water-based composition