Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Phoenix™ Safe Racer 2.0 Deluxe Kit Components
SCI_ED

Catalog No. S395193SR Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Alnico magnet
Battery holder
Binder Clip
D battery
Elastic cord
Nail
Plastic egg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S395193SR Plastic egg
S395243SR Elastic cord
S395273SR Binder Clip
S395283SR Nail
S395293SR Alnico magnet
S395313SR D battery
S395323SR Battery holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S395193SR
Add to Cart
Add to Cart

Replacement parts for Educational Kit

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.

Specifications

Color Assorted
Grade K to 5
For Use With (Application) Education Kit
Length (English) 2.5 in.
Product Type Plastic egg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.