Learn More
Med Vet International Safe-Seal Endotracheal Tube - Canine Medium-Large, J0628D, Each
Catalog No. 5000582498
Encompass_Preferred
SKU: J0628D Tube Size: Med/Large Fits (lbs): 40-60 I.D.: 10mm O.D.: 13.0mm Length (in): 16 Optional Stylet SKU: J0149STB 10mm id 13mm od 16" L
Catalog No. 50-005-82498 Supplier Med Vet International Supplier No. JOR628D
SKU: J0628D Tube Size: Med/Large Fits (lbs): 40-60 I.D.: 10mm O.D.: 13.0mm Length (in): 16 Optional Stylet SKU: J0149STB 10mm id 13mm od 16" L