Learn More
Medisca Safe-Sense™ Coverall with Hood
SDP
Catalog No. 18048008 Shop All Medisca Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-048-008 Small
18-048-007 Medium
18-048-006 Large
18-048-009 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-048-008 Supplier Medisca Supplier No. 581902
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Sterile coveralls made of microporous polypropylene with polyethylene film feature elasticated ankles, hood, and back.
  • PPE Category III
  • Disposable, full-body, one-piece protective clothing
  • Microporous polypropylene with polyethylene film
  • Zip front with sealable flap cover
  • Elasticated ankles, hood and back
  • Elastic cuffs with thumb loop
  • Low-linting and durable
  • Sterile via gamma irradiation
  • Not made with natural rubber latex
  • Manufactured in an ISO 9001 facility
  • Performance level meets EN 13034:2005, EN ISO 13982-1:2004, EN 1149-5:2008, ISO 13688:2013 and EN ISO 14325:2004
Specifications
Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Seam Type Welded
Sterility Sterile
Resistance Chemicals, Water, Liquid, Particle