Learn More
Medisca Safe-Sense™ Coverall with Hood
SDP
Catalog No. 18048009 Shop All Medisca Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-048-009 X-Large
18-048-008 Small
18-048-007 Medium
18-048-006 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-048-009 Supplier Medisca Supplier No. 582202
Sterile coveralls made of microporous polypropylene with polyethylene film feature elasticated ankles, hood, and back.
  • PPE Category III
  • Disposable, full-body, one-piece protective clothing
  • Microporous polypropylene with polyethylene film
  • Zip front with sealable flap cover
  • Elasticated ankles, hood and back
  • Elastic cuffs with thumb loop
  • Low-linting and durable
  • Sterile via gamma irradiation
  • Not made with natural rubber latex
  • Manufactured in an ISO 9001 facility
  • Performance level meets EN 13034:2005, EN ISO 13982-1:2004, EN 1149-5:2008, ISO 13688:2013 and EN ISO 14325:2004
Specifications
Product Type Cleanroom Coveralls
Size X-Large
Gender Unisex
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Seam Type Welded
Sterility Sterile
Resistance Chemicals, Water, Liquid, Particle