Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Safe-Sense™ Gloves, 12 in.
SDP

Catalog No. 18048019 Shop All Medisca Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-048-019 Small
18-048-020 Medium
18-048-021 Large
18-048-022 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-048-019 Supplier Medisca Supplier No. 578301
Add to Cart
Add to Cart
This item is not returnable. View return policy

These single use Safe-Sense™ Gloves, 12 in. provide maximum protection for non-sterile hazardous compounding administration disposal and other related chemotherapy activities.

  • They are resistant to puncture abrasion and snag.
  • These gloves are also very commonly used for non-hazardous compounding.

Specifications

Accelerator Free Yes
Applications Healthcare, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Thickness (Metric) 0.13 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.