Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Safe-Sense™ Bouffant Caps
SDP

Catalog No. 18048012 Shop All Medisca Products
Change view
Click to view available options
Style:
Pleated
Traditional
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
18-048-012 Traditional
18-048-011 Pleated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-048-012 Supplier Medisca Supplier No. 597201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bouffants are manufactured from a strong, lightweight, and breathable spun-bounded polypropylene (SPP) fabric to protect against dirt, grime, and certain dry particulates in controlled environments.

  • Bouffant caps
  • Used for cleanroom
  • Non-sterile
  • 21 in. diameter
  • Choose from pleated or traditional style

Specifications

Product Type Cleanroom Bouffant
Size 21 in.
Material Spunbond Polypropylene
Color White
Disposable Single-use
Model 5972-01
Sterility Non-sterile
Certifications/Compliance Manufactured in an ISO 13485 facility
For Use With (Application) Controlled Environments
Style Traditional
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.