Learn More
Medisca Safe-Sense™ Lab Coat
SDP
Catalog No. 18048016 Shop All Medisca Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-048-016 Small
18-048-015 Medium
18-048-014 Large
18-048-017 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-048-016 Supplier Medisca Supplier No. 575501
Only null left
Add to Cart
Add to Cart
Non-sterile disposable lab coat with knitted collar and cuffs provides breathability and comfort.
  • Breathable and comfortable lightweight material
  • SMS nonwoven laminates combine the strength and durability of spunbond with the barrier properties of meltblown microfibers
  • 5 Snap button closures for fast and easy removal
  • Knitted collar and cuffs
  • Tri-pocket design for functionality
  • Anti-static
  • Manufactured in an ISO 9001 facility
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Static
No. of Pockets 3
Collar Type Knit
Model 5755-01