Learn More
Medisca Safe-Sense™ Lab Coat
SDP
Catalog No. 18048017 Shop All Medisca Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-048-017 X-Large
18-048-016 Small
18-048-015 Medium
18-048-014 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-048-017 Supplier Medisca Supplier No. 575801
  • $57.50
    $45.65 / Pack of 10
    Save $11.85
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Non-sterile disposable lab coat with knitted collar and cuffs provides breathability and comfort.
  • Breathable and comfortable lightweight material
  • SMS nonwoven laminates combine the strength and durability of spunbond with the barrier properties of meltblown microfibers
  • 5 Snap button closures for fast and easy removal
  • Knitted collar and cuffs
  • Tri-pocket design for functionality
  • Anti-static
  • Manufactured in an ISO 9001 facility
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Static
No. of Pockets 3
Collar Type Knit
Model 5758-01