Learn More
Apexbio Technology LLC Safe DNA Gel Stain, 1ml Cas No: NA MFCD: NA
SDP
Catalog No. NC1597299
Encompass_Preferred
Safe DNA Gel Stain is a highly sensitive stain for visualization of DNA or RNA in agarose or acrylamide gels. SYBR Safe stain is specifically formulated to be a less hazardous alternative to ethidium bromide (EB) that can be used with either blue-light or UV excitation. SYBR Safe DNA Gel Stain is a better nucleic acid staining reagent for all of your molecular biology needs. SYBR Safe DNA Gel Stain is not only better for you and the environment, but also your sample and your institution. There are other sizes available. Please inqury us for quote.
Catalog No. NC1597299 Supplier Apexbio Technology LLC Supplier No. A87431
Safe DNA Gel Stain is a highly sensitive stain for visualization of DNA or RNA in agarose or acrylamide gels. SYBR Safe stain is specifically formulated to be a less hazardous alternative to ethidium bromide (EB) that can be used with either blue-light or UV excitation. SYBR Safe DNA Gel Stain is a better nucleic acid staining reagent for all of your molecular biology needs. SYBR Safe DNA Gel Stain is not only better for you and the environment, but also your sample and your institution. There are other sizes available. Please inqury us for quote.