Learn More
StatLab™ Safe-Store Block and Slide Filing System
SDP
Catalog No. STLFS85
Change view
Click to view available options
Product Type:
Filing System
Catalog No. Product Type
STLFS85 Filing System
1 options
Catalog No. STLFS85 Supplier StatLab™ Supplier No. FS8/5
Add to Cart
Add to Cart
Unique filing system can be used as an 8-drawer system for blocks or a 4-drawer system for slides.
  • Economical interlocking corrugated construction
  • Ideal for block or slid storage
  • When used as a 4 drawer file, holds approximately 2000 slides
  • When used as an 8 drawer file, holds approximately 500 tissue blocks
  • Product contains five 8-drawer block files, fully assembled, including the outer shell
  • To use for slides, simply remove the top files
Specifications
Product Type Filing System
Color White
Holds Slides or Blocks
Quantity 5 Systems/Cs.