Learn More
Description
- Comfortable face protection
- Elastic edge and headband loop for optimal fit
- Ideal for use in dental, industrial, laboratory, research, and medical industries
Specifications
Specifications
| Size | Universal |
| Color | White |
| Disposable | Single-use |
| Latex Free | Yes |
| For Use With (Application) | Dental, Industrial, Laboratory, Research, Medical |
| Product Type | Cleanroom Beard Cover |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.