Learn More
Medicom™ SafeBasics™ Fluid Impervious Gown
Catalog No. 19910745 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium/Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-745 X-Large
19-910-744 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-745 Supplier Medicom™ Supplier No. 8024
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fluid impervious gowns are designed to protect health professionals and patients from cross-contamination. Non-ISO Rated.
  • Polyethylene coating provides fluid resistance for procedures involving the potential for cross-contamination
  • Long waist ties for non-binding comfort and adjustability
  • Knitted cuffs for added protection and comfort
  • Single-use disposable and latex-free
Specifications
Size X-Large
Product Type Isolation Gown
No. of Pockets None
Gender Unisex
Color White
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Latex Free Yes
Sleeve Style Long Sleeve
Includes 50 Gowns/Cs.
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov