Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ SafeBasics™ Procedure Earloop Face Masks

Catalog No. p-7163496 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Green
White
ASTM Level:
ASTM Level 1
ASTM Level 2
ASTM Level 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color ASTM Level
19-910-649 Blue ASTM Level 1
19-910-652 Green ASTM Level 1
19-910-648 White ASTM Level 1
19-910-654 Blue ASTM Level 2
19-910-653 White ASTM Level 2
19-910-657 Blue ASTM Level 3
19-910-656 White ASTM Level 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-910-649 Supplier Medicom™ Supplier No. 2131

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultrasonically sealed procedure face masks are soft and stretchy with latex-free earloops and an adjustable nosepiece to ensure a comfortable fit.

  • Primary protection for simple, low-risk procedures involving very light splatter, moisture, spray and spatter. Ideal for patient appointments and exams
  • Inner and outer layer constructed for basic fluid resistance, with ultrasonically sealed, soft, stretchy, latex free earloops
  • Adjustable nosepiece for custom and comfortable fit and easy breathing
  • Economic alternative for simple procedures
  • ASTM Level 1, 2 and 3 protection

Specifications

Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Blue
Fastening Type Earloops
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2299, ASTM F1862
Certifications/Compliance (Europe) EN 14683
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
For Use With (Application) Animal health, health and wellness, medical, industrial, laboratory and safety
Latex Free Yes
Model 2131
Packaging Quantity 50 Masks/Box, 10 Boxes/Cs.
Pore Size 0.1 μm
Series SafeBasics™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.