Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ SafeBasics True Fit™ Thin Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 17101060 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Color:
Blue
Pink
Chlorinated:
No
Yes
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color Chlorinated
17-101-060 X-Small Pink Yes
17-101-055 X-Small Blue No
17-101-056 Small Blue No
17-101-057 Medium Blue No
17-101-058 Large Blue No
17-101-059 X-Large Blue Yes
17-101-061 Small Pink Yes
29-636-4416 Medium Pink Yes
17-101-063 Large Pink Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 17-101-060 Supplier Medicom™ Supplier No. 1186A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

SafeBasics True Fit™ Thin Nitrile Medical Examination Gloves with Textured Fingertips. This glove provides the unrivaled strength and durability of nitrile without compromising sensitivity or comfort. Textured fingertips ensure a secure grip, even when wet. Chemo tested and fentanyl safe.

  • The unrivaled strength of nitrile delivers enhanced barrier protection and exceptional fit, while providing the tactile sensitivity necessary for the most complex procedures.
  • Designed to fit like a glove should, while providing the tactile sensitivity needed for the most complex procedures.
  • Textured fingertips for a secure grip, even when wet.

Specifications

Chlorinated Yes
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Pink
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Powdered Powder Free
Size X-Small
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.