Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SafeClear II Xylene Substitutes, PROTOCOL™, Fisher HealthCare™

Catalog No. 23044192 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044192 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044192

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nonhazardous, nonflammable clearing and deparaffinization agent; Specifically designed for use in processing and staining histological/cytological specimen; Essentially odorless with low toxicity

Chemical Identifiers

CAS 64742-47-8

Specifications

Physical Form Liquid
Packaging Poly Bottle
Color Colorless
Quantity 1 gal.
Assay Percent Range 100%
Chemical Name or Material SafeClear II Xylene Substitutes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.