Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SafeClear Xylene Substitutes, PROTOCOL™, Fisher HealthCare™

Catalog No. 23314629 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-314629 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-314629

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Nonhazardous, nonflammable clearing and deparaffinization agent Specifically designed for use in processing and staining histological/cytological specimen
  • Essentially odorless with low toxicity

Chemical Identifiers

CAS 68551-16-6
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
InChI Key SGVYKUFIHHTIFL-UHFFFAOYSA-N
Synonym nonane, 2-methyl, c9-11 isoparaffin, alkanes, c9-11-iso, 2-methylnonane, unii-382akh052v, isodecane, octane, dimethyl, iso-decane, 2-methyl nonane, c10-13 isoparaffin
PubChem CID 13379
IUPAC Name 2-methylnonane
SMILES CCCCCCCC(C)C

Specifications

Physical Form Liquid
Packaging Poly Bottle
Color Colorless
Quantity 1 gal.
Assay Percent Range Assay: 100%
Chemical Name or Material SafeClear Xylene Substitutes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.