Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ SafeCup™ III 10-Panel Urine Test Cup

Catalog No. 21279258
Change view
Click to view available options
Detectable Analytes:
Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Methamphetamine (mAMP), Cannabinoids (THC)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
21-279-258 Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Methamphetamine (mAMP), Cannabinoids (THC)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-279-258 Supplier Germaine™ Supplier No. 40515
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

10-panel Urine Test Cup is a competitive binding, lateral flow immunochromatographic assay for qualitative and simultaneous detection of one or more illegal and/or prescribed substances in the urine.

Germaine™ Laboratories SafeCup™ III, 10-panel Urine Test Cup is a competitive binding, lateral flow immunochromatographic assay for qualitative and simultaneous detection of one or more illegal and/or prescribed substances in the urine. This all-in-One Cup tests for AMP, BAR, BZO, COC, MTD, MDMA, MET, OPI, PCP, THC (amphetamine, barbiturates, benzodiazpines, cocaine, methadone, mdma, methamphetamine, opiates, phencyclidine, marijuana), includes 3 validity reagents (OX/SG/pH). 25 Cups per Case, CLIA Waived for Over-the-counter, and point-of-care.

The Total Collection, Testing, and Transport System
  • Easy to Use, Easy to Read
  • Quick Results
  • Leak Proof Cap
  • Temperature Strip Included
  • Store at Room Temperature
  • Simple procedure: collect, dip and read-results in as little as five minutes

Specifications

CE Marker No
Clia Complexity Waived
Detectable Analytes Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Methamphetamine (mAMP), Cannabinoids (THC)
Type Urine Test Cup
Disposable Single-use
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.