Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories SafeCup™ III 14 Panel

Catalog No. 23111380
Change view
Click to view available options
Quantity:
25 Cups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-380 25 Cups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-380 Supplier Germaine™ Laboratories Supplier No. 44418
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories SafeCup™ III 14 Panel Urine Test Cup is competitive binding, lateral flow immunochromatographic assays.

  • Rapidly screen for broad range of illicit and prescription drugs
  • 510(k) Cleared and CLIA-waived
  • Simple procedure: collect, dip and read-results in as little as five minutes
  • Test device remains in control of administrator
  • For qualitative and simultaneous detection of Amphetamine, Barbiturate, Buprenorphine, Benzodiazepine, Cocaine, Methadone, Ecstasy, Methamphetamines, Morphine (Opiates), Oxycodone, Phencyclidine, Propoxyphene,  Tricyclic antidepressants, Marijuana

Specifications

Content And Storage 2°C to 30°C
CE Marker Yes
Detectable Analytes Cocaine (COC), Amphetamine (AMP), Methamphetamine (mAMP), Cannabinoids (THC), Morphine (MOP), Methadone (MTD), Methylenedioxymethamphetamine (MDMA), Oxycodone (OXY), Phencyclidine (PCP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Tricyclic Antidepressants (TCA), Propoxyphene (PPX)
Type Drug Cup for Drug Dip Panels
Disposable Single-use
Quantity 25 Cups
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.