Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories SafeCup™ III Urine Test Cup

Catalog No. 23111348
Change view
Click to view available options
Detectable Analytes:
Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Cocaine (COC), Cannabinoids (THC), Methylenedioxymethamphetamine (MDMA), Methamphetamine (mAMP), Methodone (MTD), Opiates (OPI), Phencyclidine (PCP)
Amphetamine (AMP), Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Cannabinoids (THC), Methylenedioxymethamphetamine (MDMA), Methamphetamine (mAMP), Methodone (MTD), Morphine (MOP), Oxycodone (OXY), Phencyclidine (PCP)
Amphetamine (AMP), Benzodiazepines (BZO), Cocaine (COC), Cannabinoids (THC), Methamphetamine (mAMP), Opiates (OPI)
Amphetamine (AMP), Cocaine (COC), Cannabinoids (THC), Methamphetamine (mAMP), Opiates (OPI)
Amphetamine (AMP), Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Phencyclidine (PCP)
Quantity:
25 Cups
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Quantity
23-111-348 Amphetamine (AMP), Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Cannabinoids (THC), Methylenedioxymethamphetamine (MDMA), Methamphetamine (mAMP), Methodone (MTD), Morphine (MOP), Oxycodone (OXY), Phencyclidine (PCP) 25 Cups
23-111-318 Amphetamine (AMP), Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Phencyclidine (PCP) 25 Cups
23-111-319 Amphetamine (AMP), Cocaine (COC), Cannabinoids (THC), Methamphetamine (mAMP), Opiates (OPI) 25 Cups
23-111-321 Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Cocaine (COC), Cannabinoids (THC), Methylenedioxymethamphetamine (MDMA), Methamphetamine (mAMP), Methodone (MTD), Opiates (OPI), Phencyclidine (PCP) 25 Cups
23-111-320 Amphetamine (AMP), Benzodiazepines (BZO), Cocaine (COC), Cannabinoids (THC), Methamphetamine (mAMP), Opiates (OPI) 25 Cups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-111-348 Supplier Germaine™ Laboratories Supplier No. 41577
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories SafeCup™ III Urine Test Cup is a competitive binding, lateral flow immunochromatographic assay for qualitative and simultaneous detection of one or more illegal and/or prescribed substances in the urine

  • Rapidly screen for broad range of illicit and prescription drugs:
    • Amphetamine (AMP)
    • Barbituates (BAR)
    • Benzodiazepines (BZO)
    • Buprenorphine (BUP)
    • Cocaine (COC)
    • Cannabis/Marijuana (THC)
    • Ecstacy (MDMA)
    • Methamphetamine (MET)
    • Methadone (MTD)
    • Opiates/Morphine (OPI/MOP)
    • Oxycodone (OXY)
    • Phencyclidine (PCP)
  • 510(k) Cleared and CLIA-waived
  • Simple procedure: collect, dip and read-results in as little as five minutes
  • Test device remains in control of administrator

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Amphetamine (AMP), Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Cannabinoids (THC), Methylenedioxymethamphetamine (MDMA), Methamphetamine (mAMP), Methodone (MTD), Morphine (MOP), Oxycodone (OXY), Phencyclidine (PCP)
Sample Type Urine
Type SafeCup for Drug Dip Panel
Disposable Single-use
Includes 12 drug panel (12-M)
Quantity 25 Cups
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.