Learn More
PDC SafeGuard™ Direct Thermal Slide Label, 15/16 in. x 15/16 in.
Catalog No. 21235830
Change view
Click to view available options
Quantity:
7200 Labels/Roll
Catalog No. Quantity
21-235-830 7200 Labels/Roll
1 options
Catalog No. 21-235-830 Supplier PDC Supplier No. TD341516
  • $1,391.00
    $1,040.65 / Case of 7200
    Save $350.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Chemical-resistant slide labels for histology with permanent adhesion, Epic EHR compatibility, and reliable performance in labs.
  • Direct thermal printable labels remove the need for protective overlays
  • Synthetic construction resists harsh chemicals including xylene, hexane, alcohol, acetone, and staining reagents
  • 4-across layout with slit separation allows quick and efficient handling
  • Permanent adhesive prevents oozing and ensures reliable attachment
  • Compact square format enhances clarity and precision in slide labeling
  • High-volume design supports large-scale lab operations
  • 3 in. core engineered for compatibility with standard lab printers
  • Epic and EHR system compatibility helps reduce errors and improve workflow
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Direct Thermal
Product Type Slide Label
Sterility Non-sterile
Quantity 7200 Labels/Roll
Dimensions (L x W) 15/16 x 15/16 in.
Length (English) 0.9375 in.
Width (English) 0.9375 in.
Width (Metric) 23.8125 mm