Learn More
PDC SafeGuard™ Direct Thermal Slide Label, 1800 Labels/Roll, 4 Rolls/Box
Catalog No. 21235827
Change view
Click to view available options
Quantity:
1800 Labels/Roll, 4 Rolls/Box
Catalog No. Quantity
21-235-827 1800 Labels/Roll, 4 Rolls/Box
1 options
Catalog No. 21-235-827 Supplier PDC Supplier No. TD317878
  • $1,391.00
    $1,040.65 / Case of 4
    Save $350.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Chemical-resistant slide labels built for histology workflows, offering reliable adhesion and compatibility with Epic and other EHR systems.
  • Direct thermal printable slide labels eliminate the need for overlays
  • Synthetic construction resists chemicals like xylene, hexane, alcohol, and acetone
  • Permanent adhesive ensures strong and lasting attachment
  • Compact 7/8 in. x 7/8 in. size supports precise slide identification
  • 1-across format with 1800 labels per roll and 4 rolls per box for bulk efficiency
  • 3 in. core designed for use with standard laboratory printers
  • Compatible with Epic and other leading EHR systems for seamless integration
  • Engineered for histology and lab testing to minimize patient misidentification risks
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Direct Thermal
Product Type Slide Label
Sterility Non-sterile
Quantity 1800 Labels/Roll, 4 Rolls/Box
Dimensions (L x W) 7/8 x 7/8 in.
Length (English) 0.875 in.
Width (English) 0.875 in.
Width (Metric) 22.225 mm