Learn More
PDC SafeGuard™ Direct Thermal Slide Label, 7/8 in. x 7/8 in.
Catalog No. 21235831
Change view
Click to view available options
Quantity:
7200 Labels/Roll
Catalog No. Quantity
21-235-831 7200 Labels/Roll
1 options
Catalog No. 21-235-831 Supplier PDC Supplier No. TD347878
  • $1,391.00
    $1,040.65 / Case of 7200
    Save $350.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Direct thermal, chemical-resistant slide labels designed for Epic compatibility.
  • Printable with direct thermal technology, eliminating the need for overlays or etching, for efficient labeling workflows
  • Chemical-resistant synthetic material endures exposure to xylene, hexane, alcohol, acetone, and common staining reagents
  • Four-label layout enhances handling and printing productivity
  • Permanent adhesive ensures secure attachment and prevents label shifting or misidentification
  • High-contrast white surface improves readability for accurate tracking
  • Designed for compatibility with Epic and major EHR systems, supporting seamless integration
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Direct Thermal
Product Type Slide Label
Sterility Non-sterile
Quantity 7200 Labels/Roll
Dimensions (L x W) 7/8 x 7/8 in.
Length (English) 0.875 in.
Width (English) 0.875 in.
Width (Metric) 22.225 mm