Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC Healthcare™ SafeGuard™ Direct Thermal Slide Labels

Catalog No. 21403440
Change view
Click to view available options
Width (Metric):
2.22 cm
2.38 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
21-403-440 2.22 cm
21-403-441 2.38 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-440 Supplier PDC Healthcare™ Supplier No. TD117878
Add to Cart
Add to Cart

Highly chemical-resistant synthetic material with permanent adhesive

  • Direct thermal synthetic slide labels with chemical-resistant material for histology and medical applications
  • Designed to prevent barcode degradation and adhesive oozing, ensuring accurate specimen identification
  • Resistant to harsh chemicals (xylene, hexane, staining reagents, alcohol, acetone, etc.)
  • No need for overlay or etching systems; quick and easy printing with direct thermal printers
  • Compatible with Epic and other leading EHR systems
  • Packaged as 1,800 labels per roll, with 4 rolls per box

Specifications

Adhesive Type Permanent (Rubber-based)
Coating None
Color White
Finish Gloss
Format Label
Material Plastic
Printable Sides Single-side Printable
Print Technology Direct Thermal
Product Type Direct Thermal Printable Label
Sterility Non-sterile
Dimensions (L x W) 0.875 x 0.875 in.
Length (English) 0.875 in.
Width (English) 0.875 in.
Width (Metric) 2.22 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.