Learn More
PDC SafeGuard™ Direct Thermal Slide Labels, 7/8 in. x 7/8 in., 1 in. Core
Catalog No. 21235821
Change view
Click to view available options
Quantity:
3600 Labels/Roll, 2 Rolls/Box
Catalog No. Quantity
21-235-821 3600 Labels/Roll, 2 Rolls/Box
1 options
Catalog No. 21-235-821 Supplier PDC Supplier No. TD127878
  • $1,391.00
    $1,040.65 / Case of 2
    Save $350.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Chemical-resistant, permanently adhesive labels for reliable histology and lab use, preventing barcode degradation and labeling errors.
  • Chemical resistant labels for durability against harsh lab reagents
  • Permanently adhesive for secure attachment on slides and surfaces
  • Prevents barcode degradation and adhesive overflow for accurate tracking
  • Compatible with standard printers without requiring overlay or etching systems
  • Durable design suitable for histology and lab testing applications
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 3600 Labels/Roll, 2 Rolls/Box
Dimensions (L x W) 7/8 x 7/8 in.
Length (English) 0.875 in.
Width (English) 0.875 in.
Width (Metric) 22.225 mm