Learn More
PDC SafeGuard™ Direct Thermal Slide Labels, 7/8 in. x 7/8 in., 3 in. Core
Catalog No. 21235828
Change view
Click to view available options
Quantity:
3600 Labels/Roll, 2 Rolls/Box
Catalog No. Quantity
21-235-828 3600 Labels/Roll, 2 Rolls/Box
1 options
Catalog No. 21-235-828 Supplier PDC Supplier No. TD327878
  • $1,391.00
    $1,040.65 / Case of 2
    Save $350.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Durable, chemical-resistant direct thermal slide labels, 2-across format, designed for histology testing.
  • Printable with direct thermal technology, requiring no overlay for fast and efficient labeling
  • Chemical-resistant synthetic material withstands exposure to harsh reagents like xylene, hexane, alcohol, and acetone
  • Permanent adhesive ensures labels stay securely in place, reducing risk of mislabeling
  • Two-across layout supports efficient handling and printing in laboratory workflows
  • Engineered for histology and laboratory applications, protecting patient data and preventing errors
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Direct Thermal
Product Type Slide Label
Sterility Non-sterile
Quantity 3600 Labels/Roll, 2 Rolls/Box
Dimensions (L x W) 7/8 x 7/8 in.
Length (English) 0.875 in.
Width (English) 0.875 in.
Width (Metric) 22.225 mm