Learn More
Med Vet International Safe-Guard Suspension - (Fenbendazole) 1 Gallon
Catalog No. 502188603
Encompass_Preferred
Safe-Guard (fenbendazole) oral drench removes and controls Lungworms, Stomach worms (adults & 4th stage larvae), brown stomach worms, barberpole worms, intestinal worms (adult & 4th stage larvae), hookworms, thread-necked intestinal worms, bankrupt worms, and nodular worms. Treatment shows a clear advantage in dry matter intake, feedlot gain, improved health and pounds of beef produced. NOT FOR SALE IN CALIFORNIA
Catalog No. 50-218-8603 Supplier Med Vet International Supplier No. PHVSAFESUSP
Safe-Guard (fenbendazole) oral drench removes and controls Lungworms, Stomach worms (adults & 4th stage larvae), brown stomach worms, barberpole worms, intestinal worms (adult & 4th stage larvae), hookworms, thread-necked intestinal worms, bankrupt worms, and nodular worms. Treatment shows a clear advantage in dry matter intake, feedlot gain, improved health and pounds of beef produced. NOT FOR SALE IN CALIFORNIA