Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC SafeGuard™ Thermal Transfer Slide Label, 15/16 in. x 15/16 in., 7200/Roll

Catalog No. 21235881
Change view
Click to view available options
Quantity:
7200/Roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-235-881 7200/Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-235-881 Supplier PDC Supplier No. TT341516
  • $1,503.00
    $1,124.65 / Case of 7200

    Save $378.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Durable, Epic-compatible synthetic slide labels for thermal transfer printing, designed for easy handling and chemical resistance.

  • Designed for thermal transfer printing, enabling fast, accurate, and efficient labeling workflows
  • Chemical-resistant synthetic material protects against harsh reagents like xylene, hexane, alcohol, acetone, and staining chemicals
  • Prevents barcode degradation and adhesive oozing, ensuring accurate sample identification
  • Slit 4-across layout allows quick handling and precise placement in laboratory settings
  • Compatible with Epic and major EHR systems, supporting seamless integration into lab workflows

Specifications

Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Thermal Transfer
Product Type Slide Label
Sterility Non-sterile
Quantity 7200/Roll
Dimensions (L x W) 15/16 x 15/16 in.
Length (English) 0.9375 in.
Width (English) 0.9375 in.
Width (Metric) 23.8125 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.