Learn More
PDC SafeGuard™ Thermal Transfer Slide Label, 4-across, 15/16 in. x 15/16 in.
Catalog No. 21235879
Change view
Click to view available options
Quantity:
7200 Labels/Roll
Catalog No. Quantity
21-235-879 7200 Labels/Roll
1 options
Catalog No. 21-235-879 Supplier PDC Supplier No. TT141516
  • $1,503.00
    $1,124.65 / Case of 7200
    Save $378.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Chemical-resistant thermal transfer slide labels designed for histology workflows and compatible with thermal transfer printers.
  • Engineered for thermal transfer printing, providing precise, durable, and efficient labeling in laboratory workflows
  • Chemical-resistant material withstands harsh reagents including xylene, hexane, alcohol, acetone, and common staining chemicals
  • Prevents barcode degradation and adhesive oozing, ensuring accurate sample identification and workflow reliability
  • 4-across format supports easy handling and streamlined labeling in high-volume histology applications
  • Enhances laboratory productivity while maintaining consistent and reliable sample tracking
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Thermal Transfer
Product Type Slide Label
Sterility Non-sterile
Quantity 7200 Labels/Roll
Dimensions (L x W) 15/16 x 15/16 in.
Length (English) 0.9375 in.
Width (English) 0.9375 in.
Width (Metric) 23.8125 mm