Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC SafeGuard™ Thermal Transfer Slide Label, 4-across, 7/8 in. x 7/8 in.

Catalog No. 21235882
Change view
Click to view available options
Quantity:
7200/Roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-235-882 7200/Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-235-882 Supplier PDC Supplier No. TT347878
  • $1,522.00
    $1,124.65 / Case of 7200

    Save $397.35

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical-resistant thermal transfer labels for slide identification in histology labs, ensuring accurate and long-lasting barcodes.

  • Engineered for thermal transfer printing, enabling precise, durable, and efficient labeling in laboratory workflows
  • Chemical-resistant material withstands harsh reagents including xylene, alcohol, acetone, hexane, and staining chemicals
  • Prevents barcode degradation and adhesive oozing to maintain accurate sample identification
  • 4-across format allows fast handling and streamlined printing in high-volume histology applications
  • Supports consistent and reliable labeling, enhancing laboratory workflow efficiency

Specifications

Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Thermal Transfer
Product Type Slide Label
Sterility Non-sterile
Quantity 7200/Roll
Dimensions (L x W) 7/8 x 7/8 in.
Length (English) 0.875 in.
Width (English) 0.875 in.
Width (Metric) 22.225 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.