Learn More
PDC SafeGuard™ Thermal Transfer Slide Label, 7/8 in. x 7/8 in., 1 in. Core
Catalog No. 21235878
Change view
Click to view available options
Quantity:
1800/Roll, 4 Rolls/Box
Catalog No. Quantity
21-235-878 1800/Roll, 4 Rolls/Box
1 options
Catalog No. 21-235-878 Supplier PDC Supplier No. TT117878
  • $1,503.00
    $1,124.65 / Case of 4
    Save $378.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Durable synthetic thermal transfer slide labels designed for histology testing and compatible with Epic, resistant to harsh chemicals.
  • Designed for thermal transfer printing, enabling fast, accurate, and reliable labeling in laboratory workflows
  • Chemical-resistant synthetic material protects against harsh reagents including xylene, hexane, alcohol, acetone, and common staining chemicals
  • Prevents barcode degradation and adhesive oozing, ensuring accurate sample identification and data integrity
  • Single-label layout allows easy handling and efficient workflow in histology labs
  • Compatible with Epic and other major EHR systems for seamless integration and improved lab productivity
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Thermal Transfer
Product Type Slide Label
Sterility Non-sterile
Quantity 1800/Roll, 4 Rolls/Box
Dimensions (L x W) 7/8 x 7/8 in.
Length (English) 0.875 in.
Width (English) 0.875 in.
Width (Metric) 22.225 mm