Learn More
PDC SafeGuard™ Thermal Transfer Slide Label, 7/8 in. x 7/8 in., 3 in. Core
Catalog No. 21235880
Change view
Click to view available options
Quantity:
1800/Roll, 4 Rolls/Box
Catalog No. Quantity
21-235-880 1800/Roll, 4 Rolls/Box
1 options
Catalog No. 21-235-880 Supplier PDC Supplier No. TT317878
  • $1,503.00
    $1,124.65 / Case of 4
    Save $378.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Epic-compatible slide labels made from synthetic, chemical-resistant material for thermal transfer printing and high-volume histology workflows.
  • Engineered for thermal transfer printing, providing fast, accurate, and reliable labeling in lab workflows
  • Chemical-resistant synthetic material protects against harsh reagents such as xylene, hexane, alcohol, acetone, and staining chemicals
  • Prevents barcode degradation and adhesive oozing, ensuring precise sample identification
  • Single-label layout simplifies handling and improves workflow efficiency in histology labs
  • Compatible with Epic and other leading EHR systems for seamless integration and data accuracy
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Thermal Transfer
Product Type Slide Label
Sterility Non-sterile
Quantity 1800/Roll, 4 Rolls/Box
Dimensions (L x W) 7/8 x 7/8 in.
Length (English) 0.875 in.
Width (English) 0.875 in.
Width (Metric) 22.225 mm