Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC Healthcare™ SafeGuard™ Thermal Transfer Slide Labels

Catalog No. 21403443
Change view
Click to view available options
Dimensions (L x W):
0.94 x 0.94 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
21-403-443 0.94 x 0.94 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-403-443 Supplier PDC Healthcare™ Supplier No. TT147878
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical-resistant synthetic material for histology and laboratory use

  • Thermal transfer synthetic slide labels with chemical-resistant properties for histology and laboratory applications
  • 4-across format provides efficient high-volume printing for laboratory workflows
  • Resistant to harsh chemicals such as xylene, hexane, staining reagents, alcohol, acetone, and others
  • Prevents barcode degradation and adhesive oozing, ensuring reliable specimen identification
  • Requires thermal transfer ribbon for printing, enabling high durability in demanding environments
  • Packaged as 7,200 labels per roll for cost-effective bulk usage

Specifications

Adhesive Type Permanent (Rubber-based)
Coating None
Color White
Finish Gloss
Format Label
Material Plastic
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Dimensions (L x W) 0.94 x 0.94 in.
Length (English) 0.94 in.
Width (English) 0.94 in.
Width (Metric) 2.38 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.