Learn More
Med Vet International Safe-Guard Drench For Goats (Fenbendazole), 100mg/mL, 125mL
Supplier:  Med Vet International PHVSAFEGOAT
Encompass_Preferred
Fenbendazole suspension 10% (100 mg/mL) for the removal and control of stomach worms (adults): Haemonchus contortus and Teladorsagia circumcincta. Administer orally. Do not use in lactating goats. Meat withdrawal: 6 days. NOT FOR SALE IN CALIFORNIA
Catalog No. 50-218-8506