Learn More
Electron Microscopy Sciences Kodak Darkroom Safelight Filter #13, 10 x 12"
SDP
Supplier:  Electron Microscopy Sciences 7491913
Encompass_Preferred
Scientifically designed for safelight lamps. Provides maximum safe illumination, when used as directed, plus offers protection from white light when using sensitized materials. Amber, 10x12", for all KODAK EKTACOLOR*, all KODAK PANALURE, and KODAK RESISTO Rapid Pan Papers.
Color: 10 x 12"
Catalog No. 50-346-68