Learn More
Electron Microscopy Sciences Kodak Darkroom Safelight Filter #3, 8 x 10"
SDP
Supplier:  Electron Microscopy Sciences 7491803
Encompass_Preferred
Scientifically designed for safelight lamps. Provides maximum safe illumination, when used as directed, plus offers protection from white light when using sensitized materials. For some panchromatic materials.
Color: Dark green
Size: 8 x 10"
Catalog No. 50-346-73