Learn More
Electron Microscopy Sciences Kodak Darkroom Safelight Utility Lamp Bracket
SDP
Catalog No. 5026763
Encompass_Preferred
Scientifically designed for safelight lamps. Provides maximum safe illumination, when used as directed, plus offers protection from white light when using sensitized materials. Optional bracket to position Kodak Utility Safelight Lamp, Model D, to a wall, table or workbench.
Catalog No. 50-267-63 Supplier Electron Microscopy Sciences Supplier No. 7491045
Scientifically designed for safelight lamps. Provides maximum safe illumination, when used as directed, plus offers protection from white light when using sensitized materials. Optional bracket to position Kodak Utility Safelight Lamp, Model D, to a wall, table or workbench.