Learn More
Medicom™ SafeMask™ Architect Pro™ N95 Surgical Respirator
Catalog No. p-7237497 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-330 Small
17-000-332 Large
17-000-331 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-000-330 Supplier Medicom™ Supplier No. 203214
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The SafeMask™ Architect Pro™ N95 Surgical Respirator is a medical grade respirator providing optimal respiratory protection by filtering 95% of particulate aerosols free of oil.
  • Reliable respiratory protection that filters out 95% of particulate aerosols free of oil
  • High-quality materials and duckbill shape offer optimal breathability
  • Unique single split comfort band hold mask in place without pulling or irritating skin
  • Lightweight construction ensures comfort and adjustable nose piece creates a personalized fit
  • Evaluated for use in healthcare settings according to recognized standards for biocompatibility, flammability, and fluid resistance
  • Convenient 8-hour wear time
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications/Compliance AS/NZS 1716:2012, NIOSH N95
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Pouch/Duckbill
Disposable Single-use
Filter Type N95
Product Type General Purpose Particulate Respirator
Size Small
With Exhalation Valve No
Filter Efficiency 95%
Latex Free Yes
Sterility Non-sterile
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov